Please login to your online account to display your discounted pricing

Methyle4-(trifluoromethyl)nicotinate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 4-(trifluoromethyl)nicotinate
CAS 175204-82-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6F3NO2
Formula Weight 205.13g/mol
MDL Number MFCD00101568
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03951CB

 
fisher scientific company
RF03951CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Chemical Name or Material Methyl 4-(trifluoromethyl)nicotinate
CAS 175204-82-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6F3NO2
Formula Weight 205.13g/mol
MDL Number MFCD00101568
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.