Please login to your online account to display your discounted pricing

Methyle4-oxo-3-piperidinecarboxylate hydrochloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  325760010

 View more versions of this product

Catalog No. AC325760010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 193.63
Quantity 1g
Color White to cream-pink
Chemical Name or Material Methyl 4-oxo-3-piperidinecarboxylate hydrochloride
Infrared Spectrum Authentic
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C7H11NO3·HCl
MDL Number MFCD00012769
CAS 56026-52-9

Specifications

Chemical Name or Material Methyl 4-oxo-3-piperidinecarboxylate hydrochloride
Name Note 97%
CAS 56026-52-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H11NO3·HCl
Formula Weight 193.63
MDL Number MFCD00012769