Please login to your online account to display your discounted pricing

Methyle4-oxo-3-piperidinecarboxylate hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 4-oxo-3-piperidinecarboxylate hydrochloride
Name Note 97%
CAS 56026-52-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H11NO3·HCl
Formula Weight 193.63
MDL Number MFCD00012769
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC325760010

  SDS
thermo fisher scientific
325760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 193.63
Quantity 1g
Color White to cream-pink
Chemical Name or Material Methyl 4-oxo-3-piperidinecarboxylate hydrochloride
Infrared Spectrum Authentic
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C7H11NO3·HCl
MDL Number MFCD00012769
CAS 56026-52-9