Please login to your online account to display your discounted pricing

Methyle4-methoxythiophene-3-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 4-methoxythiophene-3-carboxylate
CAS 65369-22-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8O3S
Formula Weight 172.2
MDL Number MFCD00067995
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM00444CB

  SDS
fisher scientific company
KM00444CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.2
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 4-methoxythiophene-3-carboxylate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C7H8O3S
MDL Number MFCD00067995
CAS 65369-22-4