Please login to your online account to display your discounted pricing

Methyle4-methoxythiophene-3-carboxylate, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM00444CB

 View more versions of this product

Catalog No. KM00444CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.2
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 4-methoxythiophene-3-carboxylate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C7H8O3S
MDL Number MFCD00067995
CAS 65369-22-4

Specifications

Chemical Name or Material Methyl 4-methoxythiophene-3-carboxylate
CAS 65369-22-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8O3S
Formula Weight 172.2
MDL Number MFCD00067995