missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-methoxysalicylate, 98%, ACROS Organics™

Manufacturer:  Acros Organics 174270100

 View more versions of this product

Catalog No. AC174270100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyle4-methoxysalicylate
Glass bottle
98%
CH3OC6H3(OH)CO2CH3
10, 381
Methyle2-hydroxy-4-methoxybenzoate
10g
Crystalline Powder or Crystals
Authentic
≥97.5%
C9H10O4
MFCD00008424
White through Yellow
49°C to 53°C
182.18

Chemical Identifiers

98%
CH3OC6H3(OH)CO2CH3
Methyle2-hydroxy-4-methoxybenzoate
≥97.5%
C9H10O4
MFCD00008424
Methyle4-methoxysalicylate
SDS Documents