Please login to your online account to display your discounted pricing

Methyle4-methoxy-2-indolecarboxylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 4-methoxy-2-indolecarboxylate
CAS 111258-23-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H11NO3
Formula Weight 205.21
MDL Number MFCD00134300
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378470010

  SDS
thermo fisher scientific
AC378470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 205.21
Quantity 1g
Color White to slightly yellow
Melting Point 141° to 144°C
Chemical Name or Material Methyl 4-methoxy-2-indolecarboxylate
Assay Percent Range 99%
Molecular Formula C11H11NO3
MDL Number MFCD00134300
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 111258-23-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.