Please login to your online account to display your discounted pricing

Methyle4-methoxy-2-indolecarboxylate, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC378470010

 View more versions of this product

Catalog No. AC378470010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 205.21
Quantity 1g
Color White to slightly yellow
Melting Point 141° to 144°C
Chemical Name or Material Methyl 4-methoxy-2-indolecarboxylate
Assay Percent Range 99%
Molecular Formula C11H11NO3
MDL Number MFCD00134300
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 111258-23-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material Methyl 4-methoxy-2-indolecarboxylate
CAS 111258-23-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H11NO3
Formula Weight 205.21
MDL Number MFCD00134300
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents