Please login to your online account to display your discounted pricing

Methyle4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 4-formyl-2, 5-dimethyl-1H-pyrrole-3-carboxylate
CAS 175205-91-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00203870
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00008CB

  SDS
fisher scientific company
SP00008CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.19
Quantity 250mg
Assay 97%
CAS Min % 97.0
CAS 175205-91-1
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00203870
Packaging Amber glass bottle
Chemical Name or Material Methyl 4-formyl-2, 5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Formula C9H11NO3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.