Please login to your online account to display your discounted pricing

Methyle4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SP00008CB

 View more versions of this product

Catalog No. SP00008CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 181.19
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 175205-91-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 4-formyl-2, 5-dimethyl-1H-pyrrole-3-carboxylate
Packaging Amber glass bottle
Molecular Formula C9H11NO3
MDL Number MFCD00203870
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Methyl 4-formyl-2, 5-dimethyl-1H-pyrrole-3-carboxylate
CAS 175205-91-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00203870
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents