Please login to your online account to display your discounted pricing

Methyle4-fluorobenzoylacetate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC199760250

 View more versions of this product

Catalog No. AC199760250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 196.18
Quantity 25g
Color Clear colorless to dark yellow
Density 1.22g/mL
Chemical Name or Material Methyle4-fluorobenzoylacetate
Packaging Glass bottle
Refractive Index 1.5200 to 1.5220 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula FC6H4COCH2CO2CH3
Infrared Spectrum Authentic
Molecular Formula C10H9FO3
MDL Number MFCD00000355
CAS 63131-29-3

Specifications

Chemical Name or Material Methyle4-fluorobenzoylacetate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9FO3
Linear Formula FC6H4COCH2CO2CH3
Formula Weight 196.18
MDL Number MFCD00000355