missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-fluorobenzoylacetate, 99%, ACROS Organics™

Manufacturer:  Acros Organics 199760100

 View more versions of this product

Catalog No. AC199760100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyle4-fluorobenzoylacetate
Authentic
1.5200 to 1.5220 (20°C, 589nm)
10g
99%
FC6H4COCH2CO2CH3
methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6
COC(=O)CC(=O)C1=CC=C(C=C1)F
196.177
196.18
≥98.5%
1.22g/mL
Glass bottle
Colorless to Yellow
63131-29-3
C10H9FO3
MFCD00000355
HGLVYXXPRSNKQN-UHFFFAOYSA-N
methyl 3-(4-fluorophenyl)-3-oxopropanoate
579425
Liquid

Chemical Identifiers

63131-29-3
196.177
HGLVYXXPRSNKQN-UHFFFAOYSA-N
579425
COC(=O)CC(=O)C1=CC=C(C=C1)F
C10H9FO3
MFCD00000355
methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6
methyl 3-(4-fluorophenyl)-3-oxopropanoate
SDS Documents