Please login to your online account to display your discounted pricing

Methyle4-cyanobenzoate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle4-cyanobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H7NO2
Linear Formula NCC6H4CO2CH3
Formula Weight 161.16
Synonym 4-Cyanobenzoic acid methyleester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC213280250

  SDS
thermo fisher scientific
AC213280250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 161.16
Quantity 25g
Color White to off-white
Melting Point 66° to 70°C
Assay Percent Range 98%
Molecular Formula C9H7NO2
CAS 1129-35-7
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyle4-cyanobenzoate
Linear Formula NCC6H4CO2CH3
Synonym 4-Cyanobenzoic acid methyleester
Safety and Handling

GHS Signal Word: Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.