missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-cyanobenzoate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC213280250

 View more versions of this product

Catalog No. AC213280250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyle4-cyanobenzoate
66° to 70°C
1129-35-7
Crystalline Powder
98%
NCC6H4CO2CH3
4-Cyanobenzoic acid methyleester
White to off-white
25g
Authentic
98%
C9H7NO2
161.16

Chemical Identifiers

98%
NCC6H4CO2CH3
4-Cyanobenzoic acid methyleester
98%
C9H7NO2
161.16
Methyle4-cyanobenzoate

Safety and Handling

SDS & Safety Documents

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning