Please login to your online account to display your discounted pricing

Methyle4-(cyanoacetyl)benzoate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 4-(cyanoacetyl)benzoate
Assay ≥96%
Assay Percent Range 98%
Molecular Formula C11 H9 N O3
Linear Formula NCCH2COC6H4CO2CH3
Formula Weight 203.2
MDL Number MFCD00015378
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314640010

  SDS
thermo fisher scientific
314640010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96%
Physical Form Flaky Crystals
Formula Weight 203.2
Quantity 1g
Color Beige
Melting Point 170° to 174°C
Chemical Name or Material Methyl 4-(cyanoacetyl)benzoate
Assay Percent Range 98%
Linear Formula NCCH2COC6H4CO2CH3
CAS 69316-08-1
Molecular Formula C11 H9 N O3
MDL Number MFCD00015378
Packaging Glass bottle
Infrared Spectrum Authentic