Please login to your online account to display your discounted pricing

Methyle4-(cyanoacetyl)benzoate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  314640010

 View more versions of this product

Catalog No. AC314640010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96%
Physical Form Flaky Crystals
Formula Weight 203.2
Quantity 1g
Color Beige
Melting Point 170° to 174°C
Chemical Name or Material Methyl 4-(cyanoacetyl)benzoate
Assay Percent Range 98%
Linear Formula NCCH2COC6H4CO2CH3
CAS 69316-08-1
Molecular Formula C11 H9 N O3
MDL Number MFCD00015378
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl 4-(cyanoacetyl)benzoate
Assay ≥96%
Assay Percent Range 98%
Molecular Formula C11 H9 N O3
Linear Formula NCCH2COC6H4CO2CH3
Formula Weight 203.2
MDL Number MFCD00015378