Please login to your online account to display your discounted pricing

Methyle4-chlorobenzoylacetate, 97%, ACROS Organics™

$98.40 - $98.40

Chemical Identifiers

Chemical Name or Material Methyl 4-chlorobenzoylacetate
CAS 53101-00-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H9ClO3
Formula Weight 212.63
MDL Number MFCD00216520
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC425400025

  SDS
thermo fisher scientific
425400025
Glass bottle 2.5g Each for $98.40
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 212.63
Quantity 2.5g
Color Clear light yellow
Molecular Formula C10H9ClO3
CAS 53101-00-1
Packaging Glass bottle
Chemical Name or Material Methyl 4-chlorobenzoylacetate
Assay Percent Range 97%
MDL Number MFCD00216520
Infrared Spectrum Authentic