missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-chlorobenzoylacetate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 425400025

 View more versions of this product

Catalog No. AC425400025


 

Specifications Description & Specifications

Specifications

≥96.0%
Liquid After Melting
Clear light yellow
53101-00-1
Methyl 4-chlorobenzoylacetate
MFCD00216520
212.63
2.5g
C10H9ClO3
Glass bottle
97%
Authentic

Chemical Identifiers

53101-00-1
C10H9ClO3
MFCD00216520
≥96.0%
97%
212.63
Methyl 4-chlorobenzoylacetate