Learn More
Methyl 4-bromo-3-methylbenzoate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438190050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 4-bromo-3-methylbenzoate | |
Authentic | |
Glass bottle | |
5g | |
GTZTYNPAPQKIIR-UHFFFAOYSA-N | |
methyl 4-bromo-3-methylbenzoate | |
3294148 | |
95% |
148547-19-7 | |
94% min. (GC) | |
C9H9BrO2 | |
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970 | |
CC1=C(C=CC(=C1)C(=O)OC)Br | |
229.07 | |
229.07 |
Chemical Identifiers
148547-19-7 | |
229.07 | |
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970 | |
methyl 4-bromo-3-methylbenzoate |
C9H9BrO2 | |
GTZTYNPAPQKIIR-UHFFFAOYSA-N | |
3294148 | |
CC1=C(C=CC(=C1)C(=O)OC)Br |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only