missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-bromo-3-methoxythiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

CAS 110545-67-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H7BrO3S
Formula Weight 251.10
MDL Number MFCD00173933
Chemical Name or Material Methyl 4-bromo-3-methoxythiophene-2-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
GK02499EA Fisher Scientific Company
GK02499EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

110545-67-0
100.0
C7H7BrO3S
MFCD00173933
97%
96.5
97%
251.10
Methyl 4-bromo-3-methoxythiophene-2-carboxylate

Specifications

10g
100.0
97%
96.5
Amber Glass Bottle
97%
251.10
C7H7BrO3S
MFCD00173933
110545-67-0
Methyl 4-bromo-3-methoxythiophene-2-carboxylate

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.