Please login to your online account to display your discounted pricing

Methyle4-bromo-3-methoxythiophene-2-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  GK02499EA

 View more versions of this product

Catalog No. GK02499EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 10g
Assay 97%
Formula Weight 251.10
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H7BrO3S
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material Methyl 4-bromo-3-methoxythiophene-2-carboxylate
Packaging Amber Glass Bottle
MDL Number MFCD00173933
CAS 110545-67-0
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl 4-bromo-3-methoxythiophene-2-carboxylate
CAS 110545-67-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H7BrO3S
Formula Weight 251.10
MDL Number MFCD00173933
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.