Please login to your online account to display your discounted pricing

Methyle4-(aminomethyl)benzoate hydrochloride, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396780050

 View more versions of this product

Catalog No. AC396780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 201.65
Physical Form Crystalline Powder
Assay ≥96.0%
Quantity 5g
Color White
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Glass bottle
Linear Formula NH2CH2C6H4COOCH3
Infrared Spectrum Authentic
Molecular Formula C9H11NO2·ClH
Chemical Name or Material Methyl 4-(aminomethyl)benzoate hydrochloride
Assay Percent Range 97%
Health Hazard 2 GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00182671
CAS 6232-11-7
Health Hazard 1 Warning

Specifications

Chemical Name or Material Methyl 4-(aminomethyl)benzoate hydrochloride
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11NO2·ClH
Linear Formula NH2CH2C6H4COOCH3
Formula Weight 201.65
MDL Number MFCD00182671
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents