Please login to your online account to display your discounted pricing

Methyle4-amino-3-iodobenzoate, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397070250

 View more versions of this product

Catalog No. AC397070250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 277.06
Assay ≥94%
Quantity 25g
Color Beige
Melting Point 86° to 90°C
Linear Formula NH2(I)C6H3COOCH3
CAS 19718-49-1
Chemical Name or Material Methyle4-amino-3-iodobenzoate
Assay Percent Range 95%
Molecular Formula C8H8INO2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00090423
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyle4-amino-3-iodobenzoate
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H8INO2
Linear Formula NH2(I)C6H3COOCH3
Formula Weight 277.06
MDL Number MFCD00090423
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents