missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle4-amino-3-iodobenzoate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397070050

 View more versions of this product

Catalog No. AC397070050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyle4-amino-3-iodobenzoate
86° to 90°C
19718-49-1
Glass bottle
≥94%
C8H8INO2
277.06
Beige
5g
Authentic
Crystalline Powder
95%
NH2(I)C6H3COOCH3
MFCD00090423

Chemical Identifiers

95%
NH2(I)C6H3COOCH3
MFCD00090423
≥94%
C8H8INO2
277.06
Methyle4-amino-3-iodobenzoate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.