missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-amino-3-iodobenzoate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 19718-49-1
Molecular Formula C8H8INO2
Molecular Weight (g/mol) 277.061
MDL Number MFCD00090423
InChI Key MRLVFVTVXSKAMX-UHFFFAOYSA-N
Synonym 4-amino-3-iodo-benzoic acid methyl ester, 4-amino-3-iodobenzoic acid methyl ester, acmc-1c8qj, benzoic acid, 4-amino-3-iodo-, methyl ester, methyl 3-iodo-4-aminobenzoate, methyl 4-amino-3-iodo-benzoate, methyl 4-amino-3-iodobenzoat, methyl 4-amino-3-iodobenzozte, methyl4-amino-3-iodobenzoate, pubchem10432
PubChem CID 4104649
IUPAC Name methyl 4-amino-3-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397070250
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC397070250
25g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

19718-49-1
277.061
MRLVFVTVXSKAMX-UHFFFAOYSA-N
4104649
COC(=O)C1=CC(=C(C=C1)N)I
C8H8INO2
MFCD00090423
4-amino-3-iodo-benzoic acid methyl ester, 4-amino-3-iodobenzoic acid methyl ester, acmc-1c8qj, benzoic acid, 4-amino-3-iodo-, methyl ester, methyl 3-iodo-4-aminobenzoate, methyl 4-amino-3-iodo-benzoate, methyl 4-amino-3-iodobenzoat, methyl 4-amino-3-iodobenzozte, methyl4-amino-3-iodobenzoate, pubchem10432
methyl 4-amino-3-iodobenzoate

Specifications

19718-49-1
86.0°C to 91.0°C
Authentic
NH2(I)C6H3COOCH3
Glass Bottle
25g
COC(=O)C1=CC(=C(C=C1)N)I
277.061
277.06
95%
Beige
95%
C8H8INO2
MFCD00090423
4-amino-3-iodo-benzoic acid methyl ester, 4-amino-3-iodobenzoic acid methyl ester, acmc-1c8qj, benzoic acid, 4-amino-3-iodo-, methyl ester, methyl 3-iodo-4-aminobenzoate, methyl 4-amino-3-iodo-benzoate, methyl 4-amino-3-iodobenzoat, methyl 4-amino-3-iodobenzozte, methyl4-amino-3-iodobenzoate, pubchem10432
MRLVFVTVXSKAMX-UHFFFAOYSA-N
methyl 4-amino-3-iodobenzoate
4104649
Crystalline Powder
Methyl 4-amino-3-iodobenzoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications

RUO – Research Use Only