Learn More
Methyle4-amino-3-bromobenzoate, 98%, ACROS Organics™
$74.24 - $74.24
Chemical Identifiers
CAS | 106896-49-5 |
---|---|
Molecular Formula | C8H8BrNO2 |
Molecular Weight (g/mol) | 230.061 |
MDL Number | MFCD01861385 |
InChI Key | AIUWAOALZYWQBX-UHFFFAOYSA-N |
Synonym | 2-bromo-4-methoxycarbonyl aniline, 2-bromo-4-methoxycarbonylaniline, 4-amino-3-bromo-benzoic acid methyl ester, 4-amino-3-bromobenzoic acid methyl ester, acmc-2098mp, benzoic acid, 4-amino-3-bromo-, methyl ester, methyl 4-amino-3-bromo-benzoate, methyl-4-amino-3-bromobenzoate, methyl4-amino-3-bromobenzoate, timtec-bb sbb012847 |
PubChem CID | 1515280 |
IUPAC Name | methyl 4-amino-3-bromobenzoate |
SMILES | COC(=O)C1=CC(=C(C=C1)N)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC350220010
|
Thermo Fisher Scientific
350220010 |
1g | Glass Bottle |
Each for $74.24
|
|
Chemical Identifiers
106896-49-5 | |
230.061 | |
AIUWAOALZYWQBX-UHFFFAOYSA-N | |
1515280 | |
COC(=O)C1=CC(=C(C=C1)N)Br |
C8H8BrNO2 | |
MFCD01861385 | |
2-bromo-4-methoxycarbonyl aniline, 2-bromo-4-methoxycarbonylaniline, 4-amino-3-bromo-benzoic acid methyl ester, 4-amino-3-bromobenzoic acid methyl ester, acmc-2098mp, benzoic acid, 4-amino-3-bromo-, methyl ester, methyl 4-amino-3-bromo-benzoate, methyl-4-amino-3-bromobenzoate, methyl4-amino-3-bromobenzoate, timtec-bb sbb012847 | |
methyl 4-amino-3-bromobenzoate |
Specifications
Authentic | |
Brown | |
1g | |
98% | |
MFCD01861385 | |
AIUWAOALZYWQBX-UHFFFAOYSA-N | |
methyl 4-amino-3-bromobenzoate | |
1515280 | |
Crystalline Powder | |
Methyl 4-amino-3-bromobenzoate |
Glass Bottle | |
104°C to 108°C | |
106896-49-5 | |
C8H8BrNO2 | |
2-bromo-4-methoxycarbonyl aniline, 2-bromo-4-methoxycarbonylaniline, 4-amino-3-bromo-benzoic acid methyl ester, 4-amino-3-bromobenzoic acid methyl ester, acmc-2098mp, benzoic acid, 4-amino-3-bromo-, methyl ester, methyl 4-amino-3-bromo-benzoate, methyl-4-amino-3-bromobenzoate, methyl4-amino-3-bromobenzoate, timtec-bb sbb012847 | |
COC(=O)C1=CC(=C(C=C1)N)Br | |
230.061 | |
230.06 | |
≥97.5% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.