missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC292780050
Specifications
Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate | |
Glass bottle | |
87.0°C to 89.0°C | |
159660-85-2 | |
C14H18N2O2 | |
1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester | |
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N | |
246.31 | |
246.31 | |
99% |
Authentic | |
Orange | |
5g | |
98.5% min. (GC) | |
MFCD00216930 | |
HLKWMBMESNOAMS-UHFFFAOYSA-N | |
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate | |
563385 | |
Crystals or Crystalline Powder |
Chemical Identifiers
159660-85-2 | |
246.31 | |
HLKWMBMESNOAMS-UHFFFAOYSA-N | |
563385 | |
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N |
C14H18N2O2 | |
MFCD00216930 | |
1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester | |
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate |