missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 99%, ACROS Organics™
$223.91 - $223.91
Chemical Identifiers
CAS | 159660-85-2 |
---|---|
Molecular Formula | C14H18N2O2 |
Molecular Weight (g/mol) | 246.31 |
MDL Number | MFCD00216930 |
InChI Key | HLKWMBMESNOAMS-UHFFFAOYSA-N |
Synonym | 1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester |
PubChem CID | 563385 |
IUPAC Name | methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate |
SMILES | COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC292780010
|
Thermo Fisher Scientific
292780010 |
1g | Glass bottle |
Each for $223.91
|
|
AC292780050
|
Thermo Fisher Scientific
AC292780050 |
5g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
159660-85-2 | |
246.31 | |
HLKWMBMESNOAMS-UHFFFAOYSA-N | |
563385 | |
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N |
C14H18N2O2 | |
MFCD00216930 | |
1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester | |
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate |
Specifications
Authentic | |
Orange | |
1g | |
98.5% min. (GC) | |
MFCD00216930 | |
HLKWMBMESNOAMS-UHFFFAOYSA-N | |
methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate | |
563385 | |
Crystals or Crystalline Powder | |
Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate |
Glass bottle | |
87.0°C to 89.0°C | |
159660-85-2 | |
C14H18N2O2 | |
1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester | |
COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N | |
246.31 | |
246.31 | |
99% |