Please login to your online account to display your discounted pricing

Methyle3-trifluoromethylbenzoylacetate, 95%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425450100

 View more versions of this product

Catalog No. AC425450100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 246.18
Quantity 10g
Color Clear light yellow
Packaging Glass bottle
Molecular Formula C11H9F3O3
CAS 93618-66-7
Chemical Name or Material Methyl 3-trifluoromethylbenzoylacetate
Assay Percent Range 95%
Infrared Spectrum Authentic
Refractive Index 1.4864 to 1.4884 (20°C, 589nm)
MDL Number MFCD00216522

Specifications

Chemical Name or Material Methyl 3-trifluoromethylbenzoylacetate
CAS 93618-66-7
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H9F3O3
Formula Weight 246.18
MDL Number MFCD00216522