Please login to your online account to display your discounted pricing

Methyle3-methyl-4-nitrobenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
CAS 24078-21-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NO4
Formula Weight 195.17
MDL Number MFCD00085640
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00715DA

  SDS
fisher scientific company
TL00715DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.17
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
Molecular Formula C9H9NO4
MDL Number MFCD00085640
CAS 24078-21-5
Assay Percent Range 97%