Please login to your online account to display your discounted pricing

Methyle3-methyl-4-nitrobenzoate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00715DA

 View more versions of this product

Catalog No. TL00715DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 195.17
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C9H9NO4
Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
MDL Number MFCD00085640
CAS 24078-21-5
Assay Percent Range 97%

Specifications

Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
CAS 24078-21-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NO4
Formula Weight 195.17
MDL Number MFCD00085640