Learn More
Methyl 3-methoxyacrylate, 95%, ACROS Organics™
Chemical Identifiers
CAS | 34846-90-7 |
---|---|
Molecular Formula | C5H8O3 |
Molecular Weight (g/mol) | 116.13 |
MDL Number | MFCD00128206 |
InChI Key | AUTCCPQKLPMHDN-ONEGZZNKSA-N |
Synonym | 2-propenoic acid, 3-methoxy-, methyl ester, 2e, 3-methoxyacrylic acid methyl ester, e-methyl 3-methoxyacrylate, e-methyl-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, methyl 3-methoxyacrylate, methyl trans-3-methoxy acrylate, methyl trans-3-methoxyacrylate, methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate |
PubChem CID | 5323651 |
IUPAC Name | methyl (E)-3-methoxyprop-2-enoate |
SMILES | COC=CC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC330460250
|
Thermo Fisher Scientific
330460250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC330461000
|
Thermo Fisher Scientific
330461000 |
100mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC330465000
|
Thermo Fisher Scientific
AC330465000 |
500mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
34846-90-7 | |
116.13 | |
AUTCCPQKLPMHDN-ONEGZZNKSA-N | |
5323651 | |
COC=CC(=O)OC |
C5H8O3 | |
MFCD00128206 | |
2-propenoic acid, 3-methoxy-, methyl ester, 2e, 3-methoxyacrylic acid methyl ester, e-methyl 3-methoxyacrylate, e-methyl-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, methyl 3-methoxyacrylate, methyl trans-3-methoxy acrylate, methyl trans-3-methoxyacrylate, methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate | |
methyl (E)-3-methoxyprop-2-enoate |
Specifications
1.0700g/mL | |
Authentic | |
1.4490 to 1.4510 | |
1.68 mPa.s (20°C) | |
Colorless | |
25mL | |
95% | |
H3COCH=CHCOOCH3 | |
2-propenoic acid, 3-methoxy-, methyl ester, 2e, 3-methoxyacrylic acid methyl ester, e-methyl 3-methoxyacrylate, e-methyl-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, methyl 3-methoxyacrylate, methyl trans-3-methoxy acrylate, methyl trans-3-methoxyacrylate, methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate | |
COC=CC(=O)OC | |
116.13 | |
116.13 | |
95% |
65°C | |
Glass bottle | |
1.07 | |
169.0°C | |
3.0°C to 4.0°C | |
34846-90-7 | |
C5H8O3 | |
MFCD00128206 | |
AUTCCPQKLPMHDN-ONEGZZNKSA-N | |
methyl (E)-3-methoxyprop-2-enoate | |
5323651 | |
Liquid | |
Methyl 3-methoxyacrylate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning