missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle3-methoxyacrylate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 330460250

 View more versions of this product

Catalog No. AC330460250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyle3-methoxyacrylate
1.07g/mL
Glass bottle
Liquid
95%
H3COCH=CHCOOCH3
MFCD00128206
25mL
34846-90-7
Authentic
1.4490 to 1.4510 (20°C, 589nm)
≥94%
C5H8O3
116.13
Clear colorless

Chemical Identifiers

95%
H3COCH=CHCOOCH3
MFCD00128206
≥94%
C5H8O3
116.13
Methyle3-methoxyacrylate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.