Learn More
Methyl 3-formylbenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 368050050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 3-formylbenzoate | |
52178-50-4 | |
97.5% min. (GC) | |
C9H8O3 | |
5 g | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol | |
COC(=O)C1=CC=CC(=C1)C=O | |
164.2 | |
164.2 |
98% | |
48.0°C to 55.0°C | |
Glass bottle | |
MFCD00189379 | |
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j | |
UVSBCUAQEZINCQ-UHFFFAOYSA-N | |
methyl 3-formylbenzoate | |
639145 | |
98% |
Chemical Identifiers
52178-50-4 | |
164.2 | |
UVSBCUAQEZINCQ-UHFFFAOYSA-N | |
639145 | |
COC(=O)C1=CC=CC(=C1)C=O |
C9H8O3 | |
MFCD00189379 | |
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j | |
methyl 3-formylbenzoate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO – Research Use Only