Please login to your online account to display your discounted pricing

Methyle3-chlorothiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
CAS 88105-17-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5ClO2S
Formula Weight 176.62
MDL Number MFCD00068135
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK01295DA

 
fisher scientific company
GK01295DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.62
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
Assay Percent Range 97%
Molecular Formula C6H5ClO2S
MDL Number MFCD00068135
CAS 88105-17-3
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle