Please login to your online account to display your discounted pricing

Methyle3-chlorophenylacetate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371671000

 View more versions of this product

Catalog No. AC371671000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 184.51
Physical Form Liquid
Quantity 100g
Color Clear colorless to light yellow
Density 1.2g/mL
Packaging Glass bottle
MDL Number MFCD00800679
Linear Formula ClC6H4CHC(O)OCH3
Assay Percent Range 98%
Chemical Name or Material Methyle3-chlorophenylacetate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5225 to 1.5245 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C9H9ClO2
CAS 53088-68-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Methyle3-chlorophenylacetate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHC(O)OCH3
Formula Weight 184.51
MDL Number MFCD00800679
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents