Please login to your online account to display your discounted pricing

Methyle3-chlorophenylacetate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  371670250

 View more versions of this product

Catalog No. AC371670250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 184.51
Quantity 25g
Color Clear colorless to light yellow
Density 1.2g/mL
CAS 53088-68-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHC(O)OCH3
MDL Number MFCD00800679
Packaging Glass bottle
Refractive Index 1.5225 to 1.5245 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material Methyle3-chlorophenylacetate
Assay Percent Range 98%

Specifications

Chemical Name or Material Methyle3-chlorophenylacetate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHC(O)OCH3
Formula Weight 184.51
MDL Number MFCD00800679
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents