Please login to your online account to display your discounted pricing

Methyle3-chlorobenzoate, 98%, ACROS Organics™

$112.86 - $112.86

Chemical Identifiers

Chemical Name or Material Methyl 3-chlorobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7ClO2
Linear Formula ClC6H4CO2CH3
Formula Weight 170.6
MDL Number MFCD00000592
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215590250

  SDS
thermo fisher scientific
215590250
Glass bottle 25g Each for $112.86
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 170.6
Physical Form Liquid or Low Melting Solid
Quantity 25g
Color Clear colorless to yellow
Density 104g/mL
Beilstein 09, 338
Merck Index 10, 2093
CAS 2905-65-9
Linear Formula ClC6H4CO2CH3
Assay Percent Range 98%
Infrared Spectrum Authentic
Chemical Name or Material Methyl 3-chlorobenzoate
Molecular Formula C8H7ClO2
MDL Number MFCD00000592
Packaging Glass bottle
Refractive Index 1.5310 to 1.5340 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.