Please login to your online account to display your discounted pricing

Methyle3-chlorobenzoate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  215590250

 View more versions of this product

Catalog No. AC215590250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid or Low Melting Solid
Assay ≥97.5%
Formula Weight 170.6
Quantity 25g
Color Clear colorless to yellow
Density 104g/mL
Beilstein 09, 338
Merck Index 10, 2093
Chemical Name or Material Methyl 3-chlorobenzoate
Assay Percent Range 98%
Molecular Formula C8H7ClO2
Linear Formula ClC6H4CO2CH3
CAS 2905-65-9
Infrared Spectrum Authentic
Refractive Index 1.5310 to 1.5340 (20°C, 589nm)
MDL Number MFCD00000592
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

Specifications

Chemical Name or Material Methyl 3-chlorobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7ClO2
Linear Formula ClC6H4CO2CH3
Formula Weight 170.6
MDL Number MFCD00000592
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents