Please login to your online account to display your discounted pricing

Methyle3-chloro-4-methylthiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-chloro-4-methylthiophene-2-carboxylate
CAS 175137-11-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H7ClO2S
Formula Weight 190.65
MDL Number MFCD00052072
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK01297EA

 
fisher scientific company
GK01297EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.65
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Methyl 3-chloro-4-methylthiophene-2-carboxylate
CAS 175137-11-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00052072
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C7H7ClO2S