Please login to your online account to display your discounted pricing

Methyle3-amino-5-(tert-butyl)thiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
CAS 175137-03-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H15NO2S
Formula Weight 213.3
MDL Number MFCD00068163
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK00671DA

 
fisher scientific company
GK00671DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GK00671EA

 
fisher scientific company
GK00671EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.3
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Assay Percent Range 97%
Molecular Formula C10H15NO2S
MDL Number MFCD00068163
CAS 175137-03-8
CAS Min % 96.5