Please login to your online account to display your discounted pricing

Methyle3-amino-5-(tert-butyl)thiophene-2-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  GK00671EA

 View more versions of this product

Catalog No. GK00671EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 213.3
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Packaging Amber glass bottle
Molecular Formula C10H15NO2S
MDL Number MFCD00068163
CAS 175137-03-8
CAS Min % 96.5
Assay Percent Range 97%

Specifications

Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
CAS 175137-03-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H15NO2S
Formula Weight 213.3
MDL Number MFCD00068163