Learn More
Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 368280050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 3-amino-5, 6-dichloro-2-pyrazinecarboxylate | |
1458-18-0 | |
97.5% min. (HPLC) | |
C6H5Cl2N3O2 | |
5g | |
USYMCUGEGUFUBI-UHFFFAOYSA-N | |
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate | |
73828 | |
98% |
98% | |
Authentic | |
Glass bottle | |
MFCD00010431 | |
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate | |
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N | |
222.03 | |
222.03 |
Chemical Identifiers
1458-18-0 | |
222.03 | |
USYMCUGEGUFUBI-UHFFFAOYSA-N | |
73828 | |
COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N |
C6H5Cl2N3O2 | |
MFCD00010431 | |
methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate | |
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environ
GHS Signal Word: Warning
EINECSNumber : 215-947-7
RUO â Research Use Only