Please login to your online account to display your discounted pricing

Methyle3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
CAS 175137-08-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10FNO2S
Formula Weight 251.28
MDL Number MFCD00068133
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK01138EA

 
fisher scientific company
GK01138EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.28
Quantity 10g
Assay 97%
CAS Max % 100.0
MDL Number MFCD00068133
CAS 175137-08-3
CAS Min % 96.5
Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H10FNO2S