Please login to your online account to display your discounted pricing

Methyle3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  GK01138EA

 View more versions of this product

Catalog No. GK01138EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 251.28
Quantity 10g
Assay 97%
CAS Max % 100.0
MDL Number MFCD00068133
CAS 175137-08-3
CAS Min % 96.5
Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H10FNO2S

Specifications

Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
CAS 175137-08-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10FNO2S
Formula Weight 251.28
MDL Number MFCD00068133