Please login to your online account to display your discounted pricing

Methyle3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
CAS 91076-93-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNO2S
Formula Weight 267.74
MDL Number MFCD00052591
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK00598DA

 
fisher scientific company
GK00598DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.74
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Packaging Amber Glass Bottle
MDL Number MFCD00052591
CAS 91076-93-6
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Molecular Formula C12H10ClNO2S
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.