Please login to your online account to display your discounted pricing

Methyle3-amino-4-methylthiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
CAS 85006-31-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00051822
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01297DA

  SDS
fisher scientific company
SB01297DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.22
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Molecular Formula C7H9NO2S
MDL Number MFCD00051822
Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
Assay Percent Range 97%
Health Hazard 1 Warning
CAS 85006-31-1