Please login to your online account to display your discounted pricing

Methyle3-amino-4-methylthiophene-2-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01297DA

 View more versions of this product

Catalog No. SB01297DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 171.22
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Molecular Formula C7H9NO2S
MDL Number MFCD00051822
Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
Assay Percent Range 97%
Health Hazard 1 Warning
CAS 85006-31-1

Specifications

Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
CAS 85006-31-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00051822
Accessories

Warning