Please login to your online account to display your discounted pricing

Methyle3-amino-4-methylbenzoate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371660250

 View more versions of this product

Catalog No. AC371660250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 165.19
Assay ≥96.0%
Quantity 25g
Color White to cream
Melting Point 114° to 118°C
Packaging Glass bottle
Linear Formula CH3(NH)C6H3COOCH3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 18595-18-1
Chemical Name or Material Methyle3-amino-4-methylbenzoate
Assay Percent Range 97%
Molecular Formula C9H11NO2
MDL Number MFCD00025206
Infrared Spectrum Authentic
Synonym Methyl 3-amino-p-toluate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyle3-amino-4-methylbenzoate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11NO2
Linear Formula CH3(NH)C6H3COOCH3
Formula Weight 165.19
MDL Number MFCD00025206
Synonym Methyl 3-amino-p-toluate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents