Please login to your online account to display your discounted pricing

Methyle3-amino-4-methylbenzoate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle3-amino-4-methylbenzoate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11NO2
Linear Formula CH3(NH)C6H3COOCH3
Formula Weight 165.19
MDL Number MFCD00025206
Synonym Methyl 3-amino-p-toluate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC371660050

  SDS
thermo fisher scientific
AC371660050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC371660250

  SDS
thermo fisher scientific
AC371660250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 165.19
Quantity 5g
Color White to cream
Melting Point 114° to 118°C
Packaging Glass bottle
Molecular Formula C9H11NO2
Linear Formula CH3(NH)C6H3COOCH3
CAS 18595-18-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Methyle3-amino-4-methylbenzoate
Assay Percent Range 97%
MDL Number MFCD00025206
Synonym Methyl 3-amino-p-toluate
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.